Merck
  • A SnRK1-ZmRFWD3-Opaque2 Signaling Axis Regulates Diurnal Nitrogen Accumulation in Maize Seeds.
A SnRK1-ZmRFWD3-Opaque2 Signaling Axis Regulates Diurnal Nitrogen Accumulation in Maize Seeds.
The Plant cell (2020-07-24)
Chaobin Li, Weiwei Qi, Zheng Liang, Xi Yang, Zeyang Ma, Rentao Song
ABSTRACT
Zeins are the predominant storage proteins in maize (Zea mays) seeds, while Opaque2 (O2) is a master transcription factor for zein-encoding genes. How the activity of O2 is regulated and responds to external signals is yet largely unknown. Here, we show that the E3 ubiquitin ligase ZmRFWD3 interacts with O2 and positively regulates its activity by enhancing its nuclear localization. Ubiquitination of O2 enhances its interaction with maize importin1, the α-subunit of Importin-1 in maize, thus enhancing its nuclear localization ability. We further show that ZmRFWD3 can be phosphorylated by a Suc-responsive protein kinase, ZmSnRK1, which leads to its degradation. We demonstrated that the activity of O2 responds to Suc levels through the ZmSnRK1-ZmRFWD3-O2 signaling axis. Intriguingly, we found that Suc levels, as well as ZmRFWD3 levels and the cytonuclear distribution of O2, exhibit diurnal patterns in developing endosperm, leading to the diurnal transcription of O2-regulated zein genes. Loss of function in ZmRFWD3 disrupts the diurnal patterns of O2 cytonuclear distribution and zein biosynthesis, and consequently changes the C/N ratio in mature seeds. We therefore identify a SnRK1-ZmRFWD3-O2 signaling axis that transduces source-to-sink signals and coordinates C and N assimilation in developing maize seeds.
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, HRP-conjugate, 1 mg/mL, Upstate®
Pricing and availability is not currently available.