Merck
  • Faster fertilization rate in conspecific versus heterospecific matings in house mice.
Faster fertilization rate in conspecific versus heterospecific matings in house mice.
Evolution; international journal of organic evolution (2008-08-30)
Matthew D Dean, Michael W Nachman
ABSTRACT
Barriers to gene flow can arise at any stage in the reproductive sequence. Most studies of reproductive isolation focus on premating or postzygotic phenotypes, leaving the importance of differences in fertilization rate overlooked. Two closely related species of house mice, Mus domesticus and M. musculus, form a narrow hybrid zone in Europe, suggesting that one or more isolating factors operate in the face of ongoing gene flow. Here, we test for differences in fertilization rate using laboratory matings as well as in vitro sperm competition assays. In noncompetitive matings, we show that fertilization occurs significantly faster in conspecific versus heterospecific matings and that this difference arises after mating and before zygotes form. To further explore the mechanisms underlying this conspecific advantage, we used competitive in vitro assays to isolate gamete interactions. Surprisingly, we discovered that M. musculus sperm consistently outcompeted M. domesticus sperm regardless of which species donated ova. These results suggest that in vivo fertilization rate is mediated by interactions between sperm, the internal female environment, and/or contributions from male seminal fluid. We discuss the implications of faster conspecific fertilization in terms of reproductive isolation among these two naturally hybridizing species.
Sigma-Aldrich
Mineral oil, BioReagent, for molecular biology, light oil
Sigma-Aldrich
Mineral oil, meets USP testing specifications
Sigma-Aldrich
Mineral oil, suitable for preparation of Nujol mulls for infrared spectroscopy, light oil
Sigma-Aldrich
Mineral oil, PCR Reagent
Sigma-Aldrich
Hyaluronidase from bovine testes, Type VIII, lyophilized powder, 300-1,000 U/mg
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, powder, suitable for mouse embryo cell culture, 750-3000 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, lyophilized powder (essentially salt-free), 750-3000 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type I-S, lyophilized powder, 400-1000 units/mg solid
Sigma-Aldrich
Hyaluronidase from bovine testes, Type VI-S, lyophilized powder, 3,000-15,000 units/mg solid
Sigma-Aldrich
Hyaluronidase from sheep testes, Type V, lyophilized powder, ≥1,500 units/mg solid
Sigma-Aldrich
Mineral oil, light oil, suitable for mouse embryo cell culture, BioXtra
Sigma-Aldrich
Mineral oil, light
Sigma-Aldrich
Mineral oil, heavy
Sigma-Aldrich
Mineral oil, light oil, (neat)
Sigma-Aldrich
Mineral oil, BioUltra, for molecular biology
Sigma-Aldrich
Hyaluronidase from sheep testes, Type II, lyophilized powder, ≥300 units/mg solid
Sigma-Aldrich
Hyaluronidase from Streptomyces hyalurolyticus