Merck
  • Novel compound heterozygous DNAAF2 mutations cause primary ciliary dyskinesia in a Han Chinese family.
Novel compound heterozygous DNAAF2 mutations cause primary ciliary dyskinesia in a Han Chinese family.
Journal of assisted reproduction and genetics (2020-07-09)
Minghan Sun, Yi Zhang, JiyunYang, Yi Wang, Hao Tan, Hailian Wang, Tiantian Lei, Xiaojie Li, Xiaojian Zhang, Wen Xiong, Ke Dou, Yongxin Ma
ABSTRACT
Primary ciliary dyskinesia (PCD), which commonly causes male infertility, is an inherited autosomal recessive disorder. This study aimed to investigate the clinical manifestations and screen mutations associated with the dynein axonemal assembly factor 2 (DNAAF2) gene in a Han Chinese family with PCD. A three-generation family with PCD was recruited in this study. Eight family members underwent comprehensive medical examinations. Genomic DNA was extracted from the participants' peripheral blood, and targeted next-generation sequencing technology was used to perform the mutation screening. The DNAAF2 expression was analyzed by immunostaining and Western blot. The proband exhibited the typical clinical features of PCD. Spermatozoa from the proband showed complete immotility but relatively high viability. Two novel compound heterozygous mutations in the DNAAF2 gene, c.C156A [p.Y52X] and c.C26A [p.S9X], were identified. Both nonsense mutations were detected in the proband, whereas the other unaffected family members carried either none or only one of the two mutations. The two nonsense heterozygous mutations were not detected in the 600 ethnically matched normal controls or in the Genome Aggregation Database. The defect of the DNAAF2 and the outer dynein arms and inner dynein arms were notably observed in the spermatozoa from the proband by immunostaining. This study identified two novel compound heterozygous mutations of DNAAF2 leading to male infertility as a result of PCD in a Han Chinese family. The findings may enhance the understanding of the pathogenesis of PCD and improve reproductive genetic counseling in China.
Sigma-Aldrich
D.E.R. 332, used as embedding medium
Sigma-Aldrich
Anti-DNAAF2 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-DNALI1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid