Merck
  • Discovery of Protein-Protein Interaction Inhibitors by Integrating Protein Engineering and Chemical Screening Platforms.
Discovery of Protein-Protein Interaction Inhibitors by Integrating Protein Engineering and Chemical Screening Platforms.
Cell chemical biology (2020-07-30)
Timurs Maculins, Javier Garcia-Pardo, Anamarija Skenderovic, Jakob Gebel, Mateusz Putyrski, Andrew Vorobyov, Philipp Busse, Gabor Varga, Maria Kuzikov, Andrea Zaliani, Simin Rahighi, Veronique Schaeffer, Michael J Parnham, Sachdev S Sidhu, Andreas Ernst, Volker Dötsch, Masato Akutsu, Ivan Dikic
ABSTRACT
Protein-protein interactions (PPIs) govern intracellular life, and identification of PPI inhibitors is challenging. Roadblocks in assay development stemming from weak binding affinities of natural PPIs impede progress in this field. We postulated that enhancing binding affinity of natural PPIs via protein engineering will aid assay development and hit discovery. This proof-of-principle study targets PPI between linear ubiquitin chains and NEMO UBAN domain, which activates NF-κB signaling. Using phage display, we generated ubiquitin variants that bind to the functional UBAN epitope with high affinity, act as competitive inhibitors, and structurally maintain the existing PPI interface. When utilized in assay development, variants enable generation of robust cell-based assays for chemical screening. Top compounds identified using this approach directly bind to UBAN and dampen NF-κB signaling. This study illustrates advantages of integrating protein engineering and chemical screening in hit identification, a development that we anticipate will have wide application in drug discovery.
Sigma-Aldrich
TPCA-1, ≥95% (HPLC)
Supelco
Aloe-emodin, analytical standard
Sigma-Aldrich
Influenza Hemagglutinin (HA) Peptide, ≥97% (HPLC)
Millipore
Monoclonal Anti-HA−Agarose antibody produced in mouse, clone HA-7, purified immunoglobulin, PBS suspension
Sigma-Aldrich
Ubiquitin from bovine erythrocytes, BioUltra, ≥98% (SDS-PAGE), essentially salt-free, lyophilized powder