Merck
  • Inhibitory Activities of Polyphenolic Extracts of Bangladeshi Vegetables against α-Amylase, α-Glucosidase, Pancreatic Lipase, Renin, and Angiotensin-Converting Enzyme.
Inhibitory Activities of Polyphenolic Extracts of Bangladeshi Vegetables against α-Amylase, α-Glucosidase, Pancreatic Lipase, Renin, and Angiotensin-Converting Enzyme.
Foods (Basel, Switzerland) (2020-07-03)
Razia Sultana, Adeola M Alashi, Khaleda Islam, Md Saifullah, C Emdad Haque, Rotimi E Aluko
ABSTRACT
The aim of the study was to determine the in vitro enzyme inhibition activities of aqueous polyphenolic extracts of nine popular Bangladeshi vegetables, namely ash gourd, bitter gourd, brinjal, Indian spinach, kangkong, okra, ridge gourd, snake gourd, and stem amaranth. Polyphenolic glycosides were the major compounds present in the extracts. Inhibition of α-amylase (up to 100% at 1 mg/mL) was stronger than α-glucosidase inhibition (up to 70.78% at 10 mg/mL). The Indian spinach extract was the strongest inhibitor of pancreatic lipase activity (IC50 = 276.77 µg/mL), which was significantly better than that of orlistat (381.16 µg/mL), a drug. Ash gourd (76.51%), brinjal (72.48%), and snake gourd (66.82%) extracts were the most effective inhibitors of angiotensin-converting enzyme (ACE), an enzyme whose excessive activities have been associated with hypertension. Brinjal also had a significantly higher renin-inhibitory activity than the other vegetable extracts. We conclude that the vegetable extracts may have the ability to reduce enzyme activities that have been associated with hyperglycemia, hyperlipidemia, and hypertension.
Sigma-Aldrich
N-[3-(2-Furyl)acryloyl]-Phe-Gly-Gly