Merck
  • Nocodazole-Induced Expression and Phosphorylation of Anillin and Other Mitotic Proteins Are Decreased in DNA-Dependent Protein Kinase Catalytic Subunit-Deficient Cells and Rescued by Inhibition of the Anaphase-Promoting Complex/Cyclosome with proTAME but Not Apcin.
Nocodazole-Induced Expression and Phosphorylation of Anillin and Other Mitotic Proteins Are Decreased in DNA-Dependent Protein Kinase Catalytic Subunit-Deficient Cells and Rescued by Inhibition of the Anaphase-Promoting Complex/Cyclosome with proTAME but Not Apcin.
Molecular and cellular biology (2020-04-15)
Pauline Douglas, Ruiqiong Ye, Suraj Radhamani, Alexander Cobban, Nicole P Jenkins, Edward Bartlett, Jonathan Roveredo, Arminja N Kettenbach, Susan P Lees-Miller
ABSTRACT
The DNA-dependent protein kinase catalytic subunit (DNA-PKcs) has well-established roles in DNA double-strand break repair, and recently, nonrepair functions have also been reported. To better understand its cellular functions, we deleted DNA-PKcs from HeLa and A549 cells using CRISPR/Cas9. The resulting cells were radiation sensitive, had reduced expression of ataxia-telangiectasia mutated (ATM), and exhibited multiple mitotic defects. Mechanistically, nocodazole-induced upregulation of cyclin B1, anillin, and securin was decreased in DNA-PKcs-deficient cells, as were phosphorylation of Aurora A on threonine 288, phosphorylation of Polo-like kinase 1 (PLK1) on threonine 210, and phosphorylation of targeting protein for Xenopus Klp2 (TPX2) on serine 121. Moreover, reduced nocodazole-induced expression of anillin, securin, and cyclin B1 and phosphorylation of PLK1, Aurora A, and TPX2 were rescued by inhibition of the anaphase-promoting complex/cyclosome (APC/C) by proTAME, which prevents binding of the APC/C-activating proteins Cdc20 and Cdh1 to the APC/C. Altogether, our studies suggest that loss of DNA-PKcs prevents inactivation of the APC/C in nocodazole-treated cells.
Product Description
proTAME, ≥90% (HPLC)
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-ATM Antibody, clone AM9, ascites fluid, clone AM9, Upstate®
Sigma-Aldrich
Anti-phospho-Ser/Thr-Pro MPM-2 Antibody, clone MPM-2, Upstate®, from mouse
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit
Sigma-Aldrich
Anti-mTOR Antibody, clone 21A12.2, clone 21A12.2, from mouse