Merck
  • Metabolic Profiling Reveals Significant Perturbations of Intracellular Glucose Homeostasis in Enterovirus-Infected Cells.
Metabolic Profiling Reveals Significant Perturbations of Intracellular Glucose Homeostasis in Enterovirus-Infected Cells.
Metabolites (2020-07-29)
Zijiao Zou, Jessica Oi-Ling Tsang, Bingpeng Yan, Kenn Ka-Heng Chik, Chris Chun-Yiu Chan, Jianli Cao, Ronghui Liang, Kaiming Tang, Feifei Yin, Zi-Wei Ye, Hin Chu, Jasper Fuk-Woo Chan, Shuofeng Yuan, Kwok-Yung Yuen
ABSTRACT
Enterovirus A71 (EV-A71) is a common cause of hand, foot, and mouth disease. Severe EV-A71 infections may be associated with life-threatening neurological complications. However, the pathogenic mechanisms underlying these severe clinical and pathological features remain incompletely understood. Metabolites are known to play critical roles in multiple stages of the replication cycles of viruses. The metabolic reprogramming induced by viral infections is essential for optimal virus replication and may be potential antiviral targets. In this study, we applied targeted metabolomics profiling to investigate the metabolic changes of induced pluripotent human stem cell (iPSC)-derived neural progenitor cells (NPCs) upon EV-A71 infection. A targeted quantitation of polar metabolites identified 14 candidates with altered expression profiles. A pathway enrichment analysis pinpointed glucose metabolic pathways as being highly perturbed upon EV-A71 infection. Gene silencing of one of the key enzymes of glycolysis, 6-phosphofructo-2-kinase (PFKFB3), significantly suppressed EV-A71 replication in vitro. Collectively, we demonstrated the feasibility to manipulate EV-A71-triggered host metabolic reprogramming as a potential anti-EV-A71 strategy.
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Serotonin hydrochloride, powder
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Dihydroxyacetone phosphate lithium salt, ≥95.0% (TLC)
Sigma-Aldrich
Phospho(enol)pyruvic acid monosodium salt hydrate, ≥97% (enzymatic)