Merck
  • A single residue, arginine 65, is critical for the functional interaction of leukocyte-associated inhibitory receptor-1 with collagens.
A single residue, arginine 65, is critical for the functional interaction of leukocyte-associated inhibitory receptor-1 with collagens.
Journal of immunology (Baltimore, Md. : 1950) (2009-04-22)
Xiaobin Tang, Sriram Narayanan, Giovanna Peruzzi, Akintomide Apara, Kannan Natarajan, David H Margulies, John E Coligan, Francisco Borrego
ABSTRACT
ITIM-containing receptors play an essential role in modulating immune responses. Leukocyte-associated inhibitory receptor (LAIR)-1, also known as CD305, is an ITIM-containing inhibitory receptor, expressed by all leukocytes, that binds collagens. In this article, we investigate the effect of a conservative R65K mutation on LAIR-1 ligand binding and function. Compared with LAIR-1 wild-type (wt)-expressing cells, LAIR-1 R65K cells show markedly reduced binding to collagen, which correlates with a reduced level of LAIR-1 polarization to the site of interaction with collagens. Both LAIR-1 wt and R65K cells can generate intracellular signals when ligated by anti-LAIR-1 mAb, but only LAIR-1 wt cells respond to collagens or matrigel. In agreement, surface plasmon resonance analyses showed that LAIR-1 R65K protein has markedly reduced avidity for collagen type I compared with LAIR-1 wt. Likewise, LAIR-1 R65K protein has decreased avidity for cells expressing transmembrane collagen XVII. Thus, a single residue, Arg65, is critical for the interaction of LAIR-1 with collagens.
Sigma-Aldrich
Urea hydrogen peroxide, 97%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Poly-L-lysine solution, 0.1 % (w/v) in H2O
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Supelco
Hydrogen peroxide solution, 30 % (w/w), for inorganic trace analysis, for ultratrace analysis
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Poly-L-lysine solution, 0.01%, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Poly-L-lysine solution, mol wt 150,000-300,000, 0.01%, sterile-filtered, BioReagent, suitable for cell culture