Merck
  • Gelatin-based nanoparticles as drug and gene delivery systems: reviewing three decades of research.
Gelatin-based nanoparticles as drug and gene delivery systems: reviewing three decades of research.
Journal of controlled release : official journal of the Controlled Release Society (2013-10-08)
Ahmed O Elzoghby
ABSTRACT
Gelatin is one of the most versatile natural biopolymers widely used in pharmaceutical industries due to its biocompatibility, biodegradability, low cost and numerous available active groups for attaching targeting molecules. These advantages led to its application in the synthesis of nanoparticles for drug and gene delivery during the last thirty years. The current article entails a general review of the different preparation techniques of gelatin nanoparticles (GNPs): desolvation, coacervation-phase separation, emulsification-solvent evaporation, reverse phase microemulsion, nanoprecipitation, self-assembly and layer-by-layer coating, from the point of view of the methodological and mechanistic aspects involved. Various crosslinkers used to improve the physicochemical properties of GNPs includintg aldehydes, genipin, carbodiimide/N-hydroxysuccinimide, and transglutaminase are reported. An analysis is given of the physicochemical behavior of GNPs including drug loading, release, particle size, zeta-potential, cytotoxicity, cellular uptake and stability. This review also attempts to provide an overview of the major applications of GNPs in drug delivery and gene therapy and their in vivo pharmacological performances, as well as site-specific drug targeting using various ligands modifying the surface of GNPs. Finally, nanocomplexes of gelatin with polymers, lipids or inorganic materials are also discussed.
Sigma-Aldrich
mPEG functionalized gelatin, 50% PEGylation, PEG average Mn 1k, gel strength 300 g Bloom
Pricing and availability is not currently available.