Merck
  • A facile technology for the high-throughput sequencing of the paired VH:VL and TCRβ:TCRα repertoires.
A facile technology for the high-throughput sequencing of the paired VH:VL and TCRβ:TCRα repertoires.
Science advances (2020-05-20)
Hidetaka Tanno, Jonathan R McDaniel, Christopher A Stevens, William N Voss, Jie Li, Russell Durrett, Jiwon Lee, Jimmy Gollihar, Yuri Tanno, George Delidakis, Arti Pothukuchy, Jared W Ellefson, Jörg J Goronzy, Jennifer A Maynard, Andrew D Ellington, Gregory C Ippolito, George Georgiou
ABSTRACT
Natively paired sequencing (NPS) of B cell receptors [variable heavy (VH) and light (VL)] and T cell receptors (TCRb and TCRa) is essential for the understanding of adaptive immunity in health and disease. Despite many recent technical advances, determining the VH:VL or TCRb:a repertoire with high accuracy and throughput remains challenging. We discovered that the recently engineered xenopolymerase, RTX, is exceptionally resistant to cell lysate inhibition in single-cell emulsion droplets. We capitalized on the characteristics of this enzyme to develop a simple, rapid, and inexpensive in-droplet overlap extension reverse transcription polymerase chain reaction method for NPS not requiring microfluidics or other specialized equipment. Using this technique, we obtained high yields (5000 to >20,000 per sample) of paired VH:VL or TCRb:a clonotypes at low cost. As a demonstration, we performed NPS on peripheral blood plasmablasts and T follicular helper cells following seasonal influenza vaccination and discovered high-affinity influenza-specific antibodies and TCRb:a.
Sigma-Aldrich
Span® 80, nonionic surfactant
Sigma-Aldrich
Human Serum, from human male AB plasma, USA origin, sterile-filtered
Sigma-Aldrich
Ionomycin from Streptomyces conglobatus, ≥98% (HPLC)
Sigma-Aldrich
TWEEN® 80, BioXtra, viscous liquid
Sigma-Aldrich
Mineral oil, BioReagent, Molecular Biology, light oil
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Triton X-100, Molecular Biology