Merck
Molecular Evolution of Auxin-Mediated Root Initiation in Plants.
Molecular biology and evolution (2019-09-11)
Jie Yu, Yuyun Zhang, Wu Liu, Hua Wang, Shaoting Wen, Yijing Zhang, Lin Xu
ABSTRACT
The root originated independently in euphyllophytes (ferns and seed plants) and lycophytes; however, the molecular evolutionary route of root initiation remains elusive. By analyses of the fern Ceratopteris richardii and seed plants, here we show that the molecular pathway involving auxin, intermediate-clade WUSCHEL-RELATED HOMEOBOX (IC-WOX) genes, and WUSCHEL-clade WOX (WC-WOX) genes could be conserved in root initiation. We propose that the "auxin>IC-WOX>WC-WOX" module in root initiation might have arisen in the common ancestor of euphyllophytes during the second origin of roots, and that this module has further developed during the evolution of different root types in ferns and seed plants.
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Supelco
Naptalam, PESTANAL®, analytical standard
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Picloram, BioReagent, suitable for plant cell culture
Supelco
Picloram, PESTANAL®, analytical standard