Merck
  • Pimozide Alleviates Hyperglycemia in Diet-Induced Obesity by Inhibiting Skeletal Muscle Ketone Oxidation.
Pimozide Alleviates Hyperglycemia in Diet-Induced Obesity by Inhibiting Skeletal Muscle Ketone Oxidation.
Cell metabolism (2020-04-11)
Rami Al Batran, Keshav Gopal, Megan E Capozzi, Jadin J Chahade, Bruno Saleme, S Amirhossein Tabatabaei-Dakhili, Amanda A Greenwell, Jingjing Niu, Malak Almutairi, Nikole J Byrne, Grant Masson, Ryekjang Kim, Farah Eaton, Erin E Mulvihill, Léa Garneau, Andrea R Masters, Zeruesenay Desta, Carlos A Velázquez-Martínez, Céline Aguer, Peter A Crawford, Gopinath Sutendra, Jonathan E Campbell, Jason R B Dyck, John R Ussher
ABSTRACT
Perturbations in carbohydrate, lipid, and protein metabolism contribute to obesity-induced type 2 diabetes (T2D), though whether alterations in ketone body metabolism influence T2D pathology is unknown. We report here that activity of the rate-limiting enzyme for ketone body oxidation, succinyl-CoA:3-ketoacid-CoA transferase (SCOT/Oxct1), is increased in muscles of obese mice. We also found that the diphenylbutylpiperidine pimozide, which is approved to suppress tics in individuals with Tourette syndrome, is a SCOT antagonist. Pimozide treatment reversed obesity-induced hyperglycemia in mice, which was phenocopied in mice with muscle-specific Oxct1/SCOT deficiency. These actions were dependent on pyruvate dehydrogenase (PDH/Pdha1) activity, the rate-limiting enzyme of glucose oxidation, as pimozide failed to alleviate hyperglycemia in obese mice with a muscle-specific Pdha1/PDH deficiency. This work defines a fundamental contribution of enhanced ketone body oxidation to the pathology of obesity-induced T2D, while suggesting pharmacological SCOT inhibition as a new class of anti-diabetes therapy.
Sigma-Aldrich
Lidoflazine, ≥98% (HPLC), powder
Sigma-Aldrich
Lithium acetoacetate, ≥90% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human OXCT1
Sigma-Aldrich
Anti-phospho-PDHE1-A type I (Ser293) Antibody, from rabbit, purified by affinity chromatography