Merck
  • DELTEX2 C-terminal domain recognizes and recruits ADP-ribosylated proteins for ubiquitination.
DELTEX2 C-terminal domain recognizes and recruits ADP-ribosylated proteins for ubiquitination.
Science advances (2020-09-17)
Syed Feroj Ahmed, Lori Buetow, Mads Gabrielsen, Sergio Lilla, Chatrin Chatrin, Gary J Sibbet, Sara Zanivan, Danny T Huang
ABSTRACT
Cross-talk between ubiquitination and ADP-ribosylation regulates spatiotemporal recruitment of key players in many signaling pathways. The DELTEX family ubiquitin ligases (DTX1 to DTX4 and DTX3L) are characterized by a RING domain followed by a C-terminal domain (DTC) of hitherto unknown function. Here, we use two label-free mass spectrometry techniques to investigate the interactome and ubiquitinated substrates of human DTX2 and identify a large proportion of proteins associated with the DNA damage repair pathway. We show that DTX2-catalyzed ubiquitination of these interacting proteins requires PARP1/2-mediated ADP-ribosylation and depends on the DTC domain. Using a combination of structural, biochemical, and cell-based techniques, we show that the DTX2 DTC domain harbors an ADP-ribose-binding pocket and recruits poly-ADP-ribose (PAR)-modified proteins for ubiquitination. This PAR-binding property of DTC domain is conserved across the DELTEX family E3s. These findings uncover a new ADP-ribose-binding domain that facilitates PAR-dependent ubiquitination.
Sigma-Aldrich
Maleimide, 99%
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Anti-Poly ADP-ribose Antibody, clone 10H, clone 10H, from mouse
Sigma-Aldrich
Anti-Ubiquitin Rabbit pAb, liquid, Calbiochem®