Merck
  • Identification of endogenous 1-aminopyrene as a novel mediator of progressive chronic kidney disease via aryl hydrocarbon receptor activation.
Identification of endogenous 1-aminopyrene as a novel mediator of progressive chronic kidney disease via aryl hydrocarbon receptor activation.
British journal of pharmacology (2020-03-29)
Hua Miao, Gang Cao, Xia-Qing Wu, Yuan-Yuan Chen, Dan-Qian Chen, Lin Chen, Nosratola D Vaziri, Ya-Long Feng, Wei Su, Yi Gao, Shougang Zhuang, Xiao-Yong Yu, Li Zhang, Yan Guo, Ying-Yong Zhao
ABSTRACT
Increasing evidence has indicated that the high risk of cardiovascular disease in chronic kidney disease (CKD) patients cannot be sufficiently explained by classic risk factors. Based on the least absolute shrinkage and selection operator method, we identified significantly altered renal tissue metabolites during progressive CKD in a 5/6 nephrectomized rat model and in CKD patients. Six aryl-containing metabolites (ACMs) were significantly increased from Week 1 to Week 20. They were associated with the activation of aryl hydrocarbon receptor (AhR) and its target genes including CYP1A1, CYP1A2 and CYP1B1, which were further validated by molecular docking. Our study further demonstrated that AhR signalling could be activated by ACM in patients with idiopathic membranous nephropathy, diabetic nephropathy and IgA nephropathy. Most importantly, 1-aminopyrene (AP) showed strong positive and negative correlation with serum creatinine and creatinine clearance, respectively. AP significantly up-regulated the mRNA expressions of AhR and its three target genes in both mice and NRK-52E cells, while this effect was partially weakened in AhR small hairpin RNA-treated mice and NRK-52E cells. Furthermore, dietary flavonoid supplementation ameliorated CKD and renal fibrosis through partially inhibiting the AhR activity via lowering the ACM levels. The antagonistic effect of flavonoids on AhR was deeply influenced by the number and location of hydroxyl and glycosyl groups. We uncovered that endogenous AP is a novel mediator of CKD progression via AhR activation; thus, AhR might serve as a promising target for CKD treatment.
Sigma-Aldrich
1-Aminopyrene, 97%
Sigma-Aldrich
Indole-3-acetic acid sodium salt, suitable for plant cell culture, BioReagent, ≥98%