Merck
  • Characteristic Evaluation of Chameleon Luminophore Dispersed in Polymer.
Characteristic Evaluation of Chameleon Luminophore Dispersed in Polymer.
Sensors (Basel, Switzerland) (2020-05-08)
Miku Kasai, Yosuke Sugioka, Masanori Yamamoto, Takayuki Nagata, Taku Nonomura, Keisuke Asai, Yasuchika Hasegawa
ABSTRACT
A temperature-sensitive paint (TSP) using a chameleon luminophore [ Tb 0 . 99 Eu 0 . 01 ( hfa ) 3 ( dpbp ) ] n is proposed. The chameleon luminophore was dispersed in isobutyl methacrylate polymer in a toluene solvent to fix it on a sample coupon. Temperature and pressure sensitivities of the chameleon luminophore-based TSP were measured using a spectrofluorophotometer. The emission for each wavelength was confirmed to be dependent on the temperature and pressure. The temperature and pressure sensitivities of the TSP were 0.81-2.8%/K and 0.08-0.12%/kPa, respectively. Higher temperature sensitivity can be obtained using the ratio of emissions from the two lanthanide ions, Tb III and Eu III . The temperature sensitivity when using the ratio of the emission intensities at 616 nm derived from Eu III and at 545 nm derived from Tb III was 3.2%/K, which was the highest value in the present study. In addition, the pressure sensitivity for the case using the ratio of the emission intensities at 616 and 545 nm was 4 . 8 × 10 - 2 % /kPa. Higher temperature sensitivity and lower pressure sensitivity than that with a single wavelength can be achieved using the ratio of the emission intensities at the two peak wavelengths derived from Tb III and Eu III .
Sigma-Aldrich
Isobutyl methacrylate, 97%