Merck
  • Hypoxia compensates cell cycle arrest with progenitor differentiation during angiogenesis.
Hypoxia compensates cell cycle arrest with progenitor differentiation during angiogenesis.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2020-03-31)
Bárbara Acosta-Iborra, Maria Tiana, Laura Maeso-Alonso, Rosana Hernández-Sierra, Gonzalo Herranz, Andrea Santamaria, Carlos Rey, Raquel Luna, Laura Puente-Santamaria, Margarita M Marques, Maria C Marin, Luis Del Peso, Benilde Jiménez
ABSTRACT
Angiogenesis, the main mechanism that allows vascular expansion for tissue regeneration or disease progression, is often triggered by an imbalance between oxygen consumption and demand. Here, by analyzing changes in the transcriptomic profile of endothelial cells (ECs) under hypoxia we uncovered that the repression of cell cycle entry and DNA replication stand as central responses in the early adaptation of ECs to low oxygen tension. Accordingly, hypoxia imposed a restriction in S-phase in ECs that is mediated by Hypoxia-Inducible Factors. Our results indicate that the induction of angiogenesis by hypoxia in Embryoid Bodies generated from murine Stem Cells is accomplished by the compensation of decreased S-phase entry in mature ECs and differentiation of progenitor cells. This conditioning most likely allows an optimum remodeling of the vascular network. Identification of the molecular underpinnings of cell cycle arrest by hypoxia would be relevant for the design of improved strategies aimed to suppress angiogenesis in pathological contexts where hypoxia is a driver of neovascularization.
Sigma-Aldrich
Lectin from Bandeiraea simplicifolia (Griffonia simplicifolia), Isolectin B4 (BSI-B4), biotin conjugate, lyophilized powder
Sigma-Aldrich
Cell Dissociation Solution Non-enzymatic 1x, Prepared in phosphate buffered saline without calcium and magnesium, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
TWEEN® 20, Molecular Biology, viscous liquid
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Copper(II) sulfate pentahydrate, ReagentPlus®, ≥98.0%
Sigma-Aldrich
1-Thioglycerol, liquid, BioReagent, suitable for cell culture, ≥97% (titration)
Sigma-Aldrich
Ala-Gln, 200 mM, solution, sterile-filtered, Biotechnology Performance Certified
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Triton X-100, laboratory grade