Merck
  • Global Regulation of the Histone Mark H3K36me2 Underlies Epithelial Plasticity and Metastatic Progression.
Global Regulation of the Histone Mark H3K36me2 Underlies Epithelial Plasticity and Metastatic Progression.
Cancer discovery (2020-03-20)
Salina Yuan, Ramakrishnan Natesan, Francisco J Sanchez-Rivera, Jinyang Li, Natarajan V Bhanu, Taiji Yamazoe, Jeffrey H Lin, Allyson J Merrell, Yogev Sela, Stacy K Thomas, Yanqing Jiang, Jacqueline B Plesset, Emma M Miller, Junwei Shi, Benjamin A Garcia, Scott W Lowe, Irfan A Asangani, Ben Z Stanger
ABSTRACT
Epithelial plasticity, reversible modulation of a cell's epithelial and mesenchymal features, is associated with tumor metastasis and chemoresistance, leading causes of cancer mortality. Although different master transcription factors and epigenetic modifiers have been implicated in this process in various contexts, the extent to which a unifying, generalized mechanism of transcriptional regulation underlies epithelial plasticity remains largely unknown. Here, through targeted CRISPR/Cas9 screening, we discovered two histone-modifying enzymes involved in the writing and erasing of H3K36me2 that act reciprocally to regulate epithelial-to-mesenchymal identity, tumor differentiation, and metastasis. Using a lysine-to-methionine histone mutant to directly inhibit H3K36me2, we found that global modulation of the mark is a conserved mechanism underlying the mesenchymal state in various contexts. Mechanistically, regulation of H3K36me2 reprograms enhancers associated with master regulators of epithelial-to-mesenchymal state. Our results thus outline a unifying epigenome-scale mechanism by which a specific histone modification regulates cellular plasticity and metastasis in cancer. SIGNIFICANCE: Although epithelial plasticity contributes to cancer metastasis and chemoresistance, no strategies exist for pharmacologically inhibiting the process. Here, we show that global regulation of a specific histone mark, H3K36me2, is a universal epigenome-wide mechanism that underlies epithelial-to-mesenchymal transition and mesenchymal-to-epithelial transition in carcinoma cells. These results offer a new strategy for targeting epithelial plasticity in cancer.This article is highlighted in the In This Issue feature, p. 747.
Sigma-Aldrich
Anti-WHSC1/NSD2 Antibody, clone 29D1, clone 29D1, from mouse
Sigma-Aldrich
Anti-Histone H3 Antibody, K36M mutant, from rabbit, purified by affinity chromatography