Merck
  • WNT and inflammatory signaling distinguish human Fallopian tube epithelial cell populations.
WNT and inflammatory signaling distinguish human Fallopian tube epithelial cell populations.
Scientific reports (2020-06-20)
Ian M Rose, Mallikarjun Bidarimath, Alex Webster, Andrew K Godwin, Andrea Flesken-Nikitin, Alexander Yu Nikitin
ABSTRACT
Many high-grade serous carcinomas (HGSCs) likely originate in the distal region of the Fallopian tube's epithelium (TE) before metastasizing to the ovary. Unfortunately, molecular mechanisms promoting malignancy in the distal TE are obfuscated, largely due to limited primary human TE gene expression data. Here we report an in depth bioinformatic characterization of 34 primary TE mRNA-seq samples. These samples were prepared from proximal and distal TE regions of 12 normal Fallopian tubes. Samples were segregated based on their aldehyde dehydrogenase (ALDH) activity. Distal cells form organoids with higher frequency and larger size during serial organoid formation assays when compared to proximal cells. Consistent with enrichment for stem/progenitor cells, ALDH+ cells have greater WNT signaling. Comparative evaluation of proximal and distal TE cell population's shows heightened inflammatory signaling in distal differentiated (ALDH-) TE. Furthermore, comparisons of proximal and distal TE cell populations finds that the distal ALDH+ TE cells exhibit pronounced expression of gene sets characteristic of HGSC sub-types. Overall, our study indicates increased organoid forming capacity, WNT/inflammatory signaling, and HGSC signatures underlie differences between distal and proximal regions of the human TE. These findings provide the basis for further mechanistic studies of distal TE susceptibility to the malignant transformation.
Sigma-Aldrich
TGF-β RI Kinase Inhibitor VI, SB431542, InSolution, ≥97%
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid
Sigma-Aldrich
AldeRed® ALDH Detection Assay
Sigma-Aldrich
Y-27632-CAS 331752-47-7-Calbiochem, Y-27632A, CAS 331752-47-7, is a cell-permeable, reversible, inhibitor of Rho kinases (Ki = 140 nM for p160ROCK). Enhances survival & cloning efficiency of ESC without affecting their pluripotency.