Merck
Role of phosphatidylcholine during neuronal differentiation.
IUBMB life (2011-08-06)
Luciana Paoletti, Claudia Elena, Pablo Domizi, Claudia Banchio
ABSTRACT
Neuronal differentiation is characterized by neuritogenesis and neurite outgrowth, processes, which are critically dependent on membrane biosynthesis, and therefore, on the expression and regulation of enzymes involved in phospholipid biosynthesis. During the last decade a great effort was made to clarify where membrane lipids are synthesized, how the newly synthesized membrane components reach the membrane and are inserted during neuritogenesis and to elucidate the mechanism by which the supply of new membrane components is coordinated with the demand for growth. Phosphatidylcholine is the principal and essential component for mammalian membranes. This review updates the mechanism by which phosphatidylcholine biosynthesis takes place and how it is coordinately regulated during neuronal differentiation.
Product Description
Avanti
15:0 PC, 1,2-dipentadecanoyl-sn-glycero-3-phosphocholine, powder
Avanti
24:1 (Cis) PC, Avanti Research - A Croda Brand
Avanti
20:0 PC, 1,2-diarachidoyl-sn-glycero-3-phosphocholine, chloroform
Avanti
09:0 PC, 1,2-dinonanoyl-sn-glycero-3-phosphocholine, powder
Avanti
18:0-14:0 PC, 1-stearoyl-2-myristoyl-sn-glycero-3-phosphocholine, powder