Merck
  • The melanocortin signaling cAMP axis accelerates repair and reduces mutagenesis of platinum-induced DNA damage.
The melanocortin signaling cAMP axis accelerates repair and reduces mutagenesis of platinum-induced DNA damage.
Scientific reports (2017-09-17)
Stuart G Jarrett, Katharine M Carter, Brent J Shelton, John A D'Orazio
ABSTRACT
Using primary melanocytes and HEK293 cells, we found that cAMP signaling accelerates repair of bi- and mono-functional platinum-induced DNA damage. Elevating cAMP signaling either by the agonistic MC1R ligand melanocyte stimulating hormone (MSH) or by pharmacologic cAMP induction by forskolin enhanced clearance of intrastrand cisplatin-adducts in melanocytes or MC1R-transfected HEK293 cells. MC1R antagonists human beta-defensin 3 and agouti signaling protein blocked MSH- but not forskolin-mediated enhancement of platinum-induced DNA damage. cAMP-enhanced repair of cisplatin-induced DNA damage was dependent on PKA-mediated phosphorylation of ATR on S435 which promoted ATR's interaction with the key NER factor xeroderma pigmentosum A (XPA) and facilitated recruitment of an XPA-ATR-pS435 complex to sites of cisplatin DNA damage. Moreover, we developed an oligonucleotide retrieval immunoprecipitation (ORiP) assay using a novel platinated-DNA substrate to establish kinetics of ATR-pS435 and XPA's associations with cisplatin-damaged DNA. Expression of a non-phosphorylatable ATR-S435A construct or deletion of A kinase-anchoring protein 12 (AKAP12) impeded platinum adduct clearance and prevented cAMP-mediated enhancement of ATR and XPA's associations with cisplatin-damaged DNA, indicating that ATR phosphorylation at S435 is necessary for cAMP-enhanced repair of platinum-induced damage and protection against cisplatin-induced mutagenesis. These data implicate cAMP signaling as a critical regulator of genomic stability against platinum-induced mutagenesis.
Sigma-Aldrich
Phenanthriplatin, >97% (NMR)