Merck
  • Are antibiotics substandard in Lebanon? Quantification of active pharmaceutical ingredients between brand and generics of selected antibiotics.
Are antibiotics substandard in Lebanon? Quantification of active pharmaceutical ingredients between brand and generics of selected antibiotics.
BMC pharmacology & toxicology (2020-02-24)
Eva Hobeika, Joanna Farhat, Joseph Saab, Walid Hleihel, Samar Azzi-Achkouty, Georges Sili, Souheil Hallit, Pascale Salameh
ABSTRACT
In developing countries, brand-generic substitution is not based on validated scientific evidence that confirm the therapeutic equivalence of the generic to the originator. Rather, decisions are made based on the availability of generic medications. Substitution by inappropriate preparations applies to antibiotics, which may increase the risk of resistance in case of underdosing. This analytical study aims to dose and assess for the accuracy of labeling three oral antibiotic preparations, namely ciprofloxacin hydrochloride, amoxicillin trihydrate and amoxicillin trihydrate-clavulanate potassium, the active pharmaceutical ingredients (APIs) found in brand and generic tablets available on the Lebanese market. One brand and 4 generics of ciprofloxacin tablets, 3 generic amoxicillin tablets, and 1 brand and 4 generics of amoxicillin-clavulanic acid medications, were quantified, taking 2 batches of each. According to the United States Pharmacopeia (USP) guidelines, ultra-high pressure liquid chromatography was used to measure the APIs content within tablets. The USP required assay limit of the API was taken as the main comparison criteria. Out of the 5 ciprofloxacin medications tested, all 5 were out of the 2% required range, thus being substandard. For amoxicillin, all 3 medications were within the 20% range. As for amoxicillin-clavulanic acid medications, 4 out of 5 medications met the 30% required range of clavulanic acid and one exceeded the claimed amount of clavulanic acid, while all 5 met the assay limit for amoxicillin. These findings raise safety and efficacy concerns, providing solid grounds for potential correlations of antibiotic resistance/substandard antibiotics.
Sigma-Aldrich
Triethylamine, ZerO2®, ≥99%