Merck
  • Periostin Activation of Integrin Receptors on Sensory Neurons Induces Allergic Itch.
Periostin Activation of Integrin Receptors on Sensory Neurons Induces Allergic Itch.
Cell reports (2020-04-09)
Santosh K Mishra, Joshua J Wheeler, Saumitra Pitake, Huiping Ding, Changyu Jiang, Tomoki Fukuyama, Judy S Paps, Patrick Ralph, Jacob Coyne, Michelle Parkington, Jennifer DeBrecht, Lauren C Ehrhardt-Humbert, Glenn P Cruse, Wolfgang Bäumer, Ru-Rong Ji, Mei-Chuan Ko, Thierry Olivry
ABSTRACT
Chronic allergic itch is a common symptom affecting millions of people and animals, but its pathogenesis is not fully explained. Herein, we show that periostin, abundantly expressed in the skin of patients with atopic dermatitis (AD), induces itch in mice, dogs, and monkeys. We identify the integrin αVβ3 expressed on a subset of sensory neurons as the periostin receptor. Using pharmacological and genetic approaches, we inhibited the function of neuronal integrin αVβ3, which significantly reduces periostin-induced itch in mice. Furthermore, we show that the cytokine TSLP, the application of AD-causing MC903 (calcipotriol), and house dust mites all induce periostin secretion. Finally, we establish that the JAK/STAT pathway is a key regulator of periostin secretion in keratinocytes. Altogether, our results identify a TSLP-periostin reciprocal activation loop that links the skin to the spinal cord via peripheral sensory neurons, and we characterize the non-canonical functional role of an integrin in itch.
Sigma-Aldrich
Laminin from Engelbreth-Holm-Swarm murine sarcoma basement membrane, 1-2 mg/mL in Tris-buffered saline, 0.2 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti -Dinitrophenyl antibody, Mouse monoclonal, IgE isotype, ~1 mg/mL, clone SPE-7, affinity purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Calnexin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
(2-Hydroxypropyl)-γ-cyclodextrin, solid
Sigma-Aldrich
Human Periostin ELISA Kit, for cell culture supernatants, plasma, and serum samples
Sigma-Aldrich
Anti-phospho-STAT5A/B (pSer725/730) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Allyl isothiocyanate, ≥95%, FCC
Sigma-Aldrich
Capsaicin, ≥95%, from Capsicum sp.
Sigma-Aldrich
Albumin, dinitrophenyl, lyophilized powder
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)