Merck
  • Electrosprayed nanoparticles of poly(p-dioxanone-co-melphalan) macromolecular prodrugs for treatment of xenograft ovarian carcinoma.
Electrosprayed nanoparticles of poly(p-dioxanone-co-melphalan) macromolecular prodrugs for treatment of xenograft ovarian carcinoma.
Materials science & engineering. C, Materials for biological applications (2020-04-14)
Aiping Wen, Xue Mei, Chengmin Feng, Chengyi Shen, Bing Wang, Xiaoming Zhang
ABSTRACT
Ovarian cancer is considered to be the most fatal reproductive cancers. Melphalan is used to treat ovarian cancer as an intraperitoneal chemotherapy agent. However, elucidating its pharmacokinetic behavior and preparing it for administration are challenging since it undergoes spontaneous hydrolysis. In this study, melphalan is transformed into a macromolecular prodrug by copolymerizing with p-dioxanone. The hydrophobicity of copolymer chains protects melphalan from hydrolysis. Poly(p-dioxanone-co-melphalan; PDCM) is electrosprayed and converted into nanoparticles (PDCM NPs) with diameters of ~300-350 nm to facilitate its intracellular delivery. UPLC-MS and HPLC are applied to verify and monitor the release of melphalan from PDCM NPs. PDCM NPs could suppress the proliferation of SKOV-3 cells. The IC50 of 4.3% melphalan-containing PDCM-3 NP was 70 mg/L, 72 h post administration. These suppression characteristics not only affected by the degradation and then the extracellular release of melphalan from PDCM NPs, but also the uptake via phagocytosis phenomenon in SKOV-3 cells. As revealed by flow cytometry, phagocytosis is a first-order process. Once phagocytosed, PDCM NPs are digested by lysosomes, causing a rapid release of melphalan into the cytoplasm, which ultimately causes suppression of SKOV-3 cell proliferation. Finally, the in vivo antitumor effects of PDCM NPs are verified in xenograft ovarian carcinoma. After a 20-day treatment, the tumor growth rate of the PDCM-3 NP group was (266 ± 178%) which was lower than those in the free melphalan group (367 ± 150%) and control group (648 ± 149%). Besides, significant tissue necrosis and growth suppression were observed in animals administered injections of PDCM NPs. Furthermore, the in vivo tracing results of Nile red-labeled PDCM NPs demonstrated that PDCM-3 NPs might be phagocytosed by macrophages and then taken to adjacent lymph nodes, which is a way of prevention or early treatment of lymphatic metastasis of tumors.
Sigma-Aldrich
(S)-2-Aminobutane-1,4-dithiol hydrochloride, 99% (titration)
Sigma-Aldrich
Decyltrimethylammonium bromide, ≥98.0% (NT)
Sigma-Aldrich
Mechlorethamine hydrochloride, 98%
Sigma-Aldrich
Melphalan, powder