Merck
  • Crystallographic analysis of murine constitutive androstane receptor ligand-binding domain complexed with 5alpha-androst-16-en-3alpha-ol.
Crystallographic analysis of murine constitutive androstane receptor ligand-binding domain complexed with 5alpha-androst-16-en-3alpha-ol.
Acta crystallographica. Section F, Structural biology and crystallization communications (2006-03-02)
Jeremy Vincent, Li Shan, Ming Fan, Joseph S Brunzelle, Barry M Forman, Elias J Fernandez
ABSTRACT
The constitutive androstane receptor (CAR) is a member of the nuclear receptor superfamily. In contrast to classical nuclear receptors, which possess small-molecule ligand-inducible activity, CAR exhibits constitutive transcriptional activity in the apparent absence of ligand. CAR is among the most important transcription factors; it coordinately regulates the expression of microsomal cytochrome P450 genes and other drug-metabolizing enzymes. The murine CAR ligand-binding domain (LBD) was coexpressed with the steroid receptor coactivator protein (SRC-1) receptor-interacting domain (RID) in Escherichia coli. The mCAR LBD subunit was purified away from SRC-1 by affinity, anion-exchange and size-exclusion chromatography, crystallized with androstenol and the structure of the complex determined by molecular replacement.
Sigma-Aldrich
5α-Androst-16-en-3α-ol