Merck
  • Iron chelators inhibit amyloid-β-induced production of lipocalin 2 in cultured astrocytes.
Iron chelators inhibit amyloid-β-induced production of lipocalin 2 in cultured astrocytes.
Neurochemistry international (2019-11-25)
Doortje W Dekens, Peter P De Deyn, Friederike Sap, Ulrich L M Eisel, Petrus J W Naudé
ABSTRACT
Lipocalin 2 (Lcn2) has been implicated to play a role in various neurodegenerative diseases, and normalizing its overexpression may be of therapeutic potential. Iron chelators were found to reduce Lcn2 levels in certain animal models of CNS injury. Focusing on Alzheimer's disease (AD), we found that the iron chelators deferoxamine and deferiprone inhibited amyloid-β (Aβ)-induced Lcn2 production in cultured primary astrocytes. Accordingly, Aβ-exposure increased astrocytic ferritin production, indicating the possibility that Aβ induces iron accumulation in astrocytes. This effect was not significantly modulated by Lcn2. Known neuroprotective effects of iron chelators may rely in part on normalization of Lcn2 levels.
Sigma-Aldrich
Deferoxamine mesylate salt, powder, ≥92.5% (TLC)
Sigma-Aldrich
Ammonium tetrathiomolybdate, 99.97% trace metals basis