Merck
  • Phenotypic Plasticity of Invasive Edge Glioma Stem-like Cells in Response to Ionizing Radiation.
Phenotypic Plasticity of Invasive Edge Glioma Stem-like Cells in Response to Ionizing Radiation.
Cell reports (2019-02-14)
Mutsuko Minata, Alessandra Audia, Junfeng Shi, Songjian Lu, Joshua Bernstock, Marat S Pavlyukov, Arvid Das, Sung-Hak Kim, Yong Jae Shin, Yeri Lee, Harim Koo, Kirti Snigdha, Indrayani Waghmare, Xing Guo, Ahmed Mohyeldin, Daniel Gallego-Perez, Jia Wang, Dongquan Chen, Peng Cheng, Farah Mukheef, Minerva Contreras, Joel F Reyes, Brian Vaillant, Erik P Sulman, Shi-Yuan Cheng, James M Markert, Bakhos A Tannous, Xinghua Lu, Madhuri Kango-Singh, L James Lee, Do-Hyun Nam, Ichiro Nakano, Krishna P Bhat
ABSTRACT
Unresectable glioblastoma (GBM) cells in the invading tumor edge can act as seeds for recurrence. The molecular and phenotypic properties of these cells remain elusive. Here, we report that the invading edge and tumor core have two distinct types of glioma stem-like cells (GSCs) that resemble proneural (PN) and mesenchymal (MES) subtypes, respectively. Upon exposure to ionizing radiation (IR), GSCs, initially enriched for a CD133+ PN signature, transition to a CD109+ MES subtype in a C/EBP-β-dependent manner. Our gene expression analysis of paired cohorts of patients with primary and recurrent GBMs identified a CD133-to-CD109 shift in tumors with an MES recurrence. Patient-derived CD133-/CD109+ cells are highly enriched with clonogenic, tumor-initiating, and radiation-resistant properties, and silencing CD109 significantly inhibits these phenotypes. We also report a conserved regulation of YAP/TAZ pathways by CD109 that could be a therapeutic target in GBM.
Sigma-Aldrich
Anti-Mitochondria Antibody, surface of intact mitochondria, clone 113-1, clone 113-1, Chemicon®, from mouse
Sigma-Aldrich
Anti-Olig-2 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-RalDH3 (ALDH1A3), from rabbit, purified by affinity chromatography