Merck
  • Early preclinical detection of prions in the skin of prion-infected animals.
Early preclinical detection of prions in the skin of prion-infected animals.
Nature communications (2019-01-18)
Zerui Wang, Matteo Manca, Aaron Foutz, Manuel V Camacho, Gregory J Raymond, Brent Race, Christina D Orru, Jue Yuan, Pingping Shen, Baiya Li, Yue Lang, Johnny Dang, Alise Adornato, Katie Williams, Nicholas R Maurer, Pierluigi Gambetti, Bin Xu, Witold Surewicz, Robert B Petersen, Xiaoping Dong, Brian S Appleby, Byron Caughey, Li Cui, Qingzhong Kong, Wen-Quan Zou
ABSTRACT
A definitive pre-mortem diagnosis of prion disease depends on brain biopsy for prion detection currently and no validated alternative preclinical diagnostic tests have been reported to date. To determine the feasibility of using skin for preclinical diagnosis, here we report ultrasensitive serial protein misfolding cyclic amplification (sPMCA) and real-time quaking-induced conversion (RT-QuIC) assays of skin samples from hamsters and humanized transgenic mice (Tg40h) at different time points after intracerebral inoculation with 263K and sCJDMM1 prions, respectively. sPMCA detects skin PrPSc as early as 2 weeks post inoculation (wpi) in hamsters and 4 wpi in Tg40h mice; RT-QuIC assay reveals earliest skin prion-seeding activity at 3 wpi in hamsters and 20 wpi in Tg40h mice. Unlike 263K-inoculated animals, mock-inoculated animals show detectable skin/brain PrPSc only after long cohabitation periods with scrapie-infected animals. Our study provides the proof-of-concept evidence that skin prions could be a biomarker for preclinical diagnosis of prion disease.
Sigma-Aldrich
Anti-Prion Protein Antibody, a.a. 109-112, clone 3F4, clone 3F4, Chemicon®, from mouse
Sigma-Aldrich
Sheep Anti-Mouse IgG Antibody, Species absorbed Antibody, HRP conjugate, from sheep, peroxidase conjugate