Merck
  • Is the existence of a mutualistic relationship between Lobesia botrana and Botrytis cinerea well-founded?
Is the existence of a mutualistic relationship between Lobesia botrana and Botrytis cinerea well-founded?
Archives of insect biochemistry and physiology (2020-01-19)
Fatemeh Kiaeian Moosavi, Elena Cargnus, Emanuela Torelli, Renzo Bortolomeazzi, Pietro Zandigiacomo, Francesco Pavan
ABSTRACT
Some studies have highlighted benefits for Lobesia botrana by adding Botrytis cinerea mycelium to an artificial larval diet and have suggested a mutualistic relationship between the two organisms on grapevine, hypothesizing that fungal sterols were the nutritional factor involved. Because the nutritional quality of an artificial diet should be similar to grapes to allow extrapolation of the results to the field conditions, in the current study L. botrana larval performance was compared when larvae were fed on grapes (berries) or two artificial diets either with or without enrichment with B. cinerea. Based on sterol analysis, the two artificial diets had high cholesterol content, but relative to berries showed comparable and low phytosterol contents, respectively (high- and low-phytosterol, HPh, and LPh). While larval fitness on the HPh diet was similar to berries, the LPh diet led to higher mortality and worse larval performance. The addition of the fungus compensated for the shortage in the LPh diet but did not improve the HPh diet. Supplementing the LPh diet with linoleic acid, which is supplied also from B. cinerea, partially improved larval performance. In a field experiment, females did not show any egg-laying preferences towards naturally botrytized bunches. The positive effect of B. cinerea on the moth's next generation that is reported in the literature could be a consequence of fungus developed inside berry tunnels bored by larvae. Therefore, based on our data and previous reports the existence of a mutualistic relationship between L. botrana and B. cinerea is not well-founded.
Sigma-Aldrich
Agar, microbiology tested, suitable for plant cell culture, suitable for cell culture, powder
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
α-Cellulose, powder
Sigma-Aldrich
Yeast Extract, for use in microbial growth medium
Sigma-Aldrich
Vanderzant vitamin mixture for insects, Animal-component free, BioReagent, suitable for insect cell culture
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Casein from bovine milk, technical grade
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra