Merck
  • Chromatin Decondensation by FOXP2 Promotes Human Neuron Maturation and Expression of Neurodevelopmental Disease Genes.
Chromatin Decondensation by FOXP2 Promotes Human Neuron Maturation and Expression of Neurodevelopmental Disease Genes.
Cell reports (2019-05-09)
Stephanie L Hickey, Stefano Berto, Genevieve Konopka
ABSTRACT
Forkhead box P2 (FOXP2) is a transcription factor expressed in the human brain that peaks during fetal development, and disruption in its ability to regulate downstream target genes leads to vulnerability to neurodevelopmental disorders. However, the mechanisms by which FOXP2 exerts regulatory control over targets during neuronal maturation have not been fully elucidated. Here, we use genome-wide chromatin accessibility assays and transcriptome-wide expression analyses in differentiating human neurons to show that FOXP2 represses proliferation-promoting genes in a DNA-binding-dependent manner. In contrast, FOXP2 and its cofactors, NFIA and NFIB, activate neuronal maturation genes in a manner that does not require FOXP2 to interact with DNA directly. Moreover, comparisons with expression data from the developing human brain suggest that FOXP2 and NFIA- or NFIB-dependent chromatin alterations drive maturation of excitatory cortical neurons. Thus, FOXP2 and its NFI cofactors may be specifically important for the development of cortical circuits underlying neurodevelopmental disorders.
Sigma-Aldrich
Anti-NFIB antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Forskolin, Coleus forskohlii
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Rabbit
Sigma-Aldrich
Anti-NFIA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)