Merck
  • Comparative analyses of long non-coding RNA profiles in vivo in cystic fibrosis lung airway and parenchyma tissues.
Comparative analyses of long non-coding RNA profiles in vivo in cystic fibrosis lung airway and parenchyma tissues.
Respiratory research (2019-12-18)
Parameet Kumar, Chaitali Sen, Kathryn Peters, Raymond A Frizzell, Roopa Biswas
ABSTRACT
Recent advances in the functional analyses of endogenous non-coding RNA (ncRNA) molecules, including long non-coding RNAs (LncRNAs), have provided a new perspective on the crucial roles of RNA in gene regulation. Consequently, LncRNA deregulation is a key factor in various diseases, including pulmonary disorders like Cystic Fibrosis (CF). CF is the most common life limiting recessive disease in the U.S., and is due to mutations in the CFTR gene. CF mutations, of which the most common is F508del-CFTR, prevents correct folding, trafficking and function of the mutant CFTR protein and is further manifested by the hyper-expression of pro-inflammatory cytokines and chemokines into the airway lumen leading to bronchiectasis and culminating in lung destruction. Here we report a distinct LncRNA signature and corresponding mRNAs that distinguishes CF lung (airway and parenchyma) tissues from matched non-CF controls (n = 4 each group), generated by microarray specific for LncRNAs which includes corresponding mRNA expressions. In silico analyses of the cellular processes that are impacted by these LncRNAs was performed using Gene Ontology (GO). A selected subset of LncRNAs were validated by quantitative real-time PCR. We have identified 636 LncRNAs differentially expressed in CF airway epithelium and 1974 in CF lung parenchyma compared to matched non-CF controls (fold change ≥2, p < 0.05), majority of which (> 50%) are intergenic. Interestingly, 15 of these differentially expressed LncRNAs and 9 coding mRNAs are common to airway and parenchyma tissues. GO analyses indicates that signaling pathways and cell membrane functions are significantly affected by the alteration in LncRNA expressions in CF lung tissues. Seven of the differentially expressed LncRNAs, exhibit similar expression trends in CFBE41o- compared to control cells. Understanding the mechanisms by which these LncRNAs regulate CF disease phenotype will help develop novel therapeutic targets for CF and related pulmonary diseases, such as COPD and Asthma.
Sigma-Aldrich
Minimum Essential Medium Eagle, With Earle′s salts and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Trypsin-EDTA solution, 1 ×, sterile; sterile-filtered, BioReagent, suitable for cell culture, 0.5 g porcine trypsin and 0.2 g EDTA, 4Na per liter of Hanks′ Balanced Salt Solution with phenol red
Sigma-Aldrich
CFBE41o- Human CF Bronchial Epithelial Cell Line, CFBE41o- human CF bronchial epithelial cell line was derived from a cystic fibrosis patient homozygous for the ΔF508 CFTR mutation.