Merck
  • Nucleolar activity and CENP-C regulate CENP-A and CAL1 availability for centromere assembly in meiosis.
Nucleolar activity and CENP-C regulate CENP-A and CAL1 availability for centromere assembly in meiosis.
Development (Cambridge, England) (2016-04-21)
Lucretia Kwenda, Caitriona M Collins, Anna A Dattoli, Elaine M Dunleavy
ABSTRACT
The centromere-specific histone CENP-A is the key epigenetic determinant of centromere identity. Whereas most histones are removed from mature sperm, CENP-A is retained to mark paternal centromeres. In Drosophila males we show that the centromere assembly factors CAL1 and CENP-C are required for meiotic chromosome segregation, CENP-A assembly and maintenance on sperm, as well as fertility. In meiosis, CENP-A accumulates with CAL1 in nucleoli. Furthermore, we show that CENP-C normally limits the release of CAL1 and CENP-A from nucleoli for proper centromere assembly in meiotic prophase I. Finally, we show that RNA polymerase I transcription is required for efficient CENP-A assembly in meiosis, as well as centromere tethering to nucleoli.
Sigma-Aldrich
ChIPAb+ Histone H3 (CT) - ChIP Validated Antibody and Primer Set, serum, from rabbit