- Fast analysis of recombinant monoclonal antibodies using IdeS proteolytic digestion and electrospray mass spectrometry.
Fast analysis of recombinant monoclonal antibodies using IdeS proteolytic digestion and electrospray mass spectrometry.
Analytical biochemistry (2011-05-21)
Guillaume Chevreux, Nolwenn Tilly, Nicolas Bihoreau
PMID21596014
ABSTRACT
We describe a fast and informative method to investigate the posttranslational modifications of monoclonal antibodies (MAbs). The MAb is first digested by a specific enzyme that cleaves heavy chains under the hinge domain. After reduction of disulfide bridges, three polypeptide chains of approximately 25 kDa are released and analyzed by liquid chromatography-mass spectrometry (LC-MS). By bisecting the heavy chains prior to MS analysis, this method provides a better MS resolution and facilitates the study of the N-linked glycans as well as of other modifications (loss of C-terminal lysine, pyroglutamination, and oxidation). The sample preparation and analysis can be performed within few hours.