Merck
  • EMILIN1 represents a major stromal element determining human trophoblast invasion of the uterine wall.
EMILIN1 represents a major stromal element determining human trophoblast invasion of the uterine wall.
Journal of cell science (2006-11-01)
Paola Spessotto, Roberta Bulla, Carla Danussi, Oriano Radillo, Marta Cervi, Giada Monami, Fleur Bossi, Francesco Tedesco, Roberto Doliana, Alfonso Colombatti
ABSTRACT
The detection of EMILIN1, a connective tissue glycoprotein associated with elastic fibers, at the level of the ectoplacental cone and trophoblast giant cells of developing mouse embryos (Braghetta et al., 2002) favored the idea of a structural as well as a functional role for this protein in the process of placentation. During the establishment of human placenta, a highly migratory subpopulation of extravillous trophoblasts (EVT), originating from anchoring chorionic villi, penetrate and invade the uterine wall. In this study we show that EMILIN1, produced by decidual stromal and smooth muscle uterine cells, is expressed in the stroma and in some instances as a gradient of increasing concentration in the perivascular region of modified vessels. This distribution pattern is consistent with the haptotactic directional migration observed in in vitro functional studies of freshly isolated EVT and of the immortalized HTR-8/SVneo cell line of trophoblasts. Function-blocking monoclonal antibodies against alpha4-integrin chain and against EMILIN1 as well as the use of EMILIN1-specific short interfering RNA confirmed that trophoblasts interact with EMILIN1 and/or its functional gC1q1 domain via alpha4beta1 integrin. Finally, membrane type I-matrix metalloproteinase (MT1-MMP) and MMP-2 were upregulated in co-cultures of trophoblast cells and stromal cells, suggesting a contributing role in the haptotactic process towards EMILIN1.
Sigma-Aldrich
Anti-Integrin α4 Antibody, clone P1H4, clone P1H4, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin α1 Antibody, clone FB12, clone FB12, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal Anti-HLA Class I Antigen antibody produced in mouse, clone W6/32, purified immunoglobulin, buffered aqueous solution