Merck
  • Long-lasting effects of neonatal dexamethasone treatment on spatial learning and hippocampal synaptic plasticity: involvement of the NMDA receptor complex.
Long-lasting effects of neonatal dexamethasone treatment on spatial learning and hippocampal synaptic plasticity: involvement of the NMDA receptor complex.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2003-03-11)
Patrick J G H Kamphuis, Fabrizio Gardoni, Amer Kamal, Gerda Croiset, Joost M Bakker, Flaminio Cattabeni, Willem Hendrik Gispen, Frank van Bel, Monica Di Luca, Victor M Wiegant
ABSTRACT
The effects of neonatal dexamethasone (DEX) treatment on spatial learning and hippocampal synaptic plasticity were investigated in adult rats. Spatial learning in reference and working memory versions of the Morris maze was impaired in DEX-treated rats. In hippocampal slices of DEX rats, long-term depression was facilitated and potentiation was impaired. Paired-pulse facilitation was normal, suggesting a postsynaptic defect as cause of the learning and plasticity deficits. Western blot analysis of hippocampal postsynaptic densities (PSD) revealed a reduction in NR2B subunit protein, whereas the abundance of the other major N-methyl-D-aspartate (NMDA) receptor subunits (NR1, NR2A), AMPA receptor subunits (GluR2/3), scaffolding proteins, and Ca2+/calmodulin-dependent protein kinase II (alphaCaMKII) were unaltered. This selective reduction in NR2B likely resulted from altered receptor assembly rather than subunit expression, because the abundance of NR2B in the homogenate and crude synaptosomal fractions was unaltered. In addition, the activity of alphaCaMKII, an NMDA receptor complex associated protein kinase, was increased in PSD of DEX rats. The results indicate that neonatal treatment with DEX causes alterations in composition and function of the hippocampal NMDA receptor complex that persist into adulthood. These alterations likely explain the deficits in hippocampal synaptic plasticity and spatial learning induced by neonatal DEX treatment.
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone PY20, clone PY20, Upstate®, from mouse
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, 4G10® Platinum, clone 4G10, Upstate®, from mouse