Merck
  • UXT-V1 facilitates the formation of MAVS antiviral signalosome on mitochondria.
UXT-V1 facilitates the formation of MAVS antiviral signalosome on mitochondria.
Journal of immunology (Baltimore, Md. : 1950) (2011-12-02)
Yuefeng Huang, Heng Liu, Rui Ge, Yi Zhou, Xiwen Lou, Chen Wang
ABSTRACT
Virus infection induces the MAVS-TNFR-associated factor (TRAF) 3 signaling axis on mitochondria. It remains to elucidate the corresponding regulatory processes. In this study, we identify UXT-V1 as a novel TRAF3-binding protein. UXT-V1 is critical for the virus-induced activation of NF-κB and IFN regulatory factor 3. Reduction of UXT-V1 impairs the induction of IFN-β and attenuates the host antiviral responses. The N-terminal TRAF-binding motif of UXT-V1 binds to the C-terminal TRAF domain of TRAF3, thus facilitating the interaction between TRAF3 and MAVS. Notably, TRAF3 and TNFR-associated death domain protein are recruited onto mitochondria upon virus infection. These translocations are blocked when knocking down UXT-V1. Thus, UXT-V1 represents a novel integral component of the MAVS signalosome on mitochondria, mediating the innate antiviral signal transduction.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Anti-Sp1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid