Merck
Intracellular calcium regulates nonsense-mediated mRNA decay.
Nature medicine (2014-07-30)
Andrew Nickless, Erin Jackson, Jayne Marasa, Patrick Nugent, Robert W Mercer, David Piwnica-Worms, Zhongsheng You
ABSTRACT
The nonsense-mediated mRNA decay (NMD) pathway selectively eliminates aberrant transcripts containing premature translation termination codons and regulates the levels of a number of physiological mRNAs. NMD modulates the clinical outcome of a variety of human diseases, including cancer and many genetic disorders, and may represent a target for therapeutic intervention. Here, we have developed a new multicolored bioluminescence-based reporter system that can specifically and effectively assay NMD in live human cells. Using this reporter system, we conducted a robust high-throughput small-molecule screen in human cells and, unpredictably, identified a group of cardiac glycosides, including ouabain and digoxin, as potent inhibitors of NMD. Cardiac glycoside-mediated effects on NMD are dependent on binding and inhibiting the sodium-potassium ATPase on the plasma membrane and subsequent elevation of intracellular calcium levels. Induction of calcium release from the endoplasmic reticulum also leads to inhibition of NMD. Thus, this study reveals intracellular calcium as a key regulator of NMD and has implications for exploiting NMD in the treatment of disease.
Sigma-Aldrich
BAPTA-AM, ≥95% (HPLC)
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Dialyzed by ultrafiltration against 0.15 M NaCl, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Ouabain octahydrate, ≥95% (HPLC), powder
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
Sigma-Aldrich
Calcium Ionophore A23187, ≥98% (TLC), powder
Supelco
Digoxin, analytical standard