Merck
  • Disease-associated mutations in Niemann-Pick type C1 alter ER calcium signaling and neuronal plasticity.
Disease-associated mutations in Niemann-Pick type C1 alter ER calcium signaling and neuronal plasticity.
The Journal of cell biology (2019-10-12)
Scott A Tiscione, Oscar Vivas, Kenneth S Ginsburg, Donald M Bers, Daniel S Ory, Luis F Santana, Rose E Dixon, Eamonn J Dickson
ABSTRACT
Niemann-Pick type C1 (NPC1) protein is essential for the transport of externally derived cholesterol from lysosomes to other organelles. Deficiency of NPC1 underlies the progressive NPC1 neurodegenerative disorder. Currently, there are no curative therapies for this fatal disease. Given the Ca2+ hypothesis of neurodegeneration, which posits that altered Ca2+ dynamics contribute to neuropathology, we tested if disease mutations in NPC1 alter Ca2+ signaling and neuronal plasticity. We determine that NPC1 inhibition or disease mutations potentiate store-operated Ca2+ entry (SOCE) due to a presenilin 1 (PSEN1)-dependent reduction in ER Ca2+ levels alongside elevated expression of the molecular SOCE components ORAI1 and STIM1. Associated with this dysfunctional Ca2+ signaling is destabilization of neuronal dendritic spines. Knockdown of PSEN1 or inhibition of the SREBP pathway restores Ca2+ homeostasis, corrects differential protein expression, reduces cholesterol accumulation, and rescues spine density. These findings highlight lysosomes as a crucial signaling platform responsible for tuning ER Ca2+ signaling, SOCE, and synaptic architecture in health and disease.
Sigma-Aldrich
Anti-STIM1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-MAP-2, from rabbit, purified by affinity chromatography
Sigma-Aldrich
1-β-D-Arabinofuranosylcytosine, Anticancer, antiviral agent that is especially effective against leukemias.
Sigma-Aldrich
Anti-Presenilin-1 Antibody, NT, clone hPS1-NT, culture supernatant, clone hPS1-NT, Chemicon®