Merck
  • Molecular recognition between guanine and cytosine-functionalized nucleolipid hybrid bilayers supported on gold (111) electrodes.
Molecular recognition between guanine and cytosine-functionalized nucleolipid hybrid bilayers supported on gold (111) electrodes.
Bioelectrochemistry (Amsterdam, Netherlands) (2020-01-26)
Julia Alvarez-Malmagro, ZhangFei Su, J Jay Leitch, Francisco Prieto, Manuela Rueda, Jacek Lipkowski
ABSTRACT
A hybrid bilayer lipid membrane (hBLM), constructed with a 1-hexadecanethiol self-assembled interior leaflet and a 1,2-dipalmitoyl-sn-glycero-3-cytidine nucleolipid exterior leaflet, was deposited at the surface of a gold (111) electrode. This system was used to investigate the molecular recognition reaction between the cytosine moieties of the lipid head group with guanine molecules in the bulk electrolyte solution. Electrochemical measurements and photon polarization modulation infrared reflection absorption spectroscopy (PMIRRAS) were employed to characterize the system and determine the extent of the molecular recognition reaction. The capacitance of the hBLM-covered gold electrode was very low (~1 μF cm-2), therefore the charge density at the gold surface was small. Changing the electrode potential had a minimal effect on the complexation between the cytosine moieties and guanine molecules due to small changes in the static electric field across the membrane. This behavior favored the formation of the guanine-cytosine complex.
Product Description
Avanti
16:0 CDP DG, Avanti Research - A Croda Brand 870510P, powder
Avanti
16:0 CDP DG, Avanti Research - A Croda Brand 870510C