Merck
  • Mitochondrial calcium exchange links metabolism with the epigenome to control cellular differentiation.
Mitochondrial calcium exchange links metabolism with the epigenome to control cellular differentiation.
Nature communications (2019-10-06)
Alyssa A Lombardi, Andrew A Gibb, Ehtesham Arif, Devin W Kolmetzky, Dhanendra Tomar, Timothy S Luongo, Pooja Jadiya, Emma K Murray, Pawel K Lorkiewicz, György Hajnóczky, Elizabeth Murphy, Zoltan P Arany, Daniel P Kelly, Kenneth B Margulies, Bradford G Hill, John W Elrod
ABSTRACT
Fibroblast to myofibroblast differentiation is crucial for the initial healing response but excessive myofibroblast activation leads to pathological fibrosis. Therefore, it is imperative to understand the mechanisms underlying myofibroblast formation. Here we report that mitochondrial calcium (mCa2+) signaling is a regulatory mechanism in myofibroblast differentiation and fibrosis. We demonstrate that fibrotic signaling alters gating of the mitochondrial calcium uniporter (mtCU) in a MICU1-dependent fashion to reduce mCa2+ uptake and induce coordinated changes in metabolism, i.e., increased glycolysis feeding anabolic pathways and glutaminolysis yielding increased α-ketoglutarate (αKG) bioavailability. mCa2+-dependent metabolic reprogramming leads to the activation of αKG-dependent histone demethylases, enhancing chromatin accessibility in loci specific to the myofibroblast gene program, resulting in differentiation. Our results uncover an important role for the mtCU beyond metabolic regulation and cell death and demonstrate that mCa2+ signaling regulates the epigenome to influence cellular differentiation.
Sigma-Aldrich
Angiotensin II human, ≥93% (HPLC), powder
Sigma-Aldrich
SIGMAFAST Protease Inhibitor Cocktail Tablets, EDTA-Free, for use in purification of Histidine-tagged proteins
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
Dimethyl 2-oxoglutarate, 96%
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Sigma-Aldrich
JIB-04, ≥98% (HPLC)