Merck
  • Experimental and theoretical evidence for molecular forces driving surface segregation in photonic colloidal assemblies.
Experimental and theoretical evidence for molecular forces driving surface segregation in photonic colloidal assemblies.
Science advances (2019-09-27)
Ming Xiao, Ziying Hu, Thomas E Gartner, Xiaozhou Yang, Weiyao Li, Arthi Jayaraman, Nathan C Gianneschi, Matthew D Shawkey, Ali Dhinojwala
ABSTRACT
Surface segregation in binary colloidal mixtures offers a simple way to control both surface and bulk properties without affecting their bulk composition. Here, we combine experiments and coarse-grained molecular dynamics (CG-MD) simulations to delineate the effects of particle chemistry and size on surface segregation in photonic colloidal assemblies from binary mixtures of melanin and silica particles of size ratio (Dlarge/Dsmall) ranging from 1.0 to ~2.2. We find that melanin and/or smaller particles segregate at the surface of micrometer-sized colloidal assemblies (supraballs) prepared by an emulsion process. Conversely, no such surface segregation occurs in films prepared by evaporative assembly. CG-MD simulations explain the experimental observations by showing that particles with the larger contact angle (melanin) are enriched at the supraball surface regardless of the relative strength of particle-interface interactions, a result with implications for the broad understanding and design of colloidal particle assemblies.
Sigma-Aldrich
1-Octanol, anhydrous, ≥99%