Merck
  • Cholesterol transport through the peroxisome-ER membrane contacts tethered by PI(4,5)P2 and extended synaptotagmins.
Cholesterol transport through the peroxisome-ER membrane contacts tethered by PI(4,5)P2 and extended synaptotagmins.
Science China. Life sciences (2019-05-31)
Jian Xiao, Jie Luo, Ao Hu, Ting Xiao, Meixin Li, Zekai Kong, Luyi Jiang, Zimu Zhou, Yacheng Liao, Chang Xie, Beibei Chu, Honghua Miao, Boliang Li, Xiongjie Shi, Bao-Liang Song
ABSTRACT
Most mammalian cells take up cholesterol from low-density lipoproteins (LDLs) via receptor-mediated endocytosis. After reaching lysosomes, LDL-derived cholesterol continues to transport to downstream organelles including the ER for specific structural and functional needs. Peroxisomes are recently found to receive cholesterol from lysosomes through lysosome-peroxisome membrane contacts. However, whether and how cholesterol is conveyed from peroxisomes to the ER remain unknown. Here, by combining high-resolution microscopic analyses and in vitro reconstitution of highly purified organelles or artificial liposomes, we demonstrate that peroxisomes form membrane contacts with the ER through the interaction between peroxisomal PI(4,5)P2 and ER-resident extended synaptotagmin-1, 2 and 3 (E-Syts). Depletion of peroxisomal PI(4,5)P2 or E-Syts markedly decreases peroxisome-ER membrane contacts and induces cholesterol accumulation in lysosomes. Furthermore, we show that cholesterol is delivered from 3H-labeled peroxisomes or PI(4,5)P2-containing liposomes to the ER in vitro, and that the presence of peroxisomes augments cholesterol transfer from lysosomes to the ER. Together, our study reveals a new cholesterol transport pathway along the lysosome-peroxisome-ER membrane contacts in the cell.
Sigma-Aldrich
Lysosome Isolation Kit, sufficient for 25 g (tissue), sufficient for 20 mL (packed cells), enrichment of lysosomes from tissues and packed cells
Sigma-Aldrich
Filipin complex from Streptomyces filipinensis, ≥70% (UV)