Merck
  • Differential immune gene response in gills, skin, and spleen of rainbow trout Oncorhynchus mykiss infected by Ichthyophthirius multifiliis.
Differential immune gene response in gills, skin, and spleen of rainbow trout Oncorhynchus mykiss infected by Ichthyophthirius multifiliis.
PloS one (2019-06-21)
Khairul Syahputra, Per W Kania, Azmi Al-Jubury, Huria Marnis, Agung Cahyo Setyawan, Kurt Buchmann
ABSTRACT
Infection of rainbow trout with the parasitic ciliate Ichthyopthirius multifiliis induces differential responses in gills, skin and spleen. A controlled experimental infection was performed and expression of immune-relevant genes in skin, gills, and spleen were recorded by qPCR at day 1 and 8 after parasite exposure. Infection induced a marked reaction involving regulation of innate and adaptive immune genes in rainbow trout at day 8 post-infection. The expression level of a total of 22 out of 24 investigated genes was significantly higher in gills compared to skin reflecting the more sensitive and delicate structure of gills. Especially pro-inflammatory cytokines IL-6, IL-17 C1, regulatory cytokines IL-4/13A, IL-10, TGFβ, complement factor C5, chemokines CK10, CK12, acute phase proteins (precerebellin, hepcidin) and immunoglobulins (IgM, IgT) displayed differential expression levels. The spleen, a central immune organ with no trace of the parasite, showed elevated expression of IgM, IgT, complement factor C5 and chemokine CK10 (compared to skin and gills directly exposed to the parasite), indicating an interaction between the infected surface sites and central immune organs. This communication could be mediated by chemokines CK10 and CK12 and cytokine IL-4/13A and may at least partly explain the establishment of a systemic response in rainbow trout against the parasite.
Product Description
Supelco
Ethyl 3-aminobenzoate methanesulfonate salt, analytical standard
Sigma-Aldrich
RNAlater®, Stabilize and protect RNA with immediate RNase inactivation