Merck
  • Combinatorial Pharyngeal Taste Coding for Feeding Avoidance in Adult Drosophila.
Combinatorial Pharyngeal Taste Coding for Feeding Avoidance in Adult Drosophila.
Cell reports (2019-10-24)
Yu-Chieh David Chen, Scarlet Jinhong Park, Ryan Matthew Joseph, William W Ja, Anupama Arun Dahanukar
ABSTRACT
Taste drives appropriate food preference and intake. In Drosophila, taste neurons are housed in both external and internal organs, but the latter have been relatively underexplored. Here, we report that Poxn mutants with a minimal taste system of pharyngeal neurons can avoid many aversive tastants, including bitter compounds, acid, and salt, suggesting that pharyngeal taste is sufficient for rejecting intake of aversive compounds. Optogenetic activation of selected pharyngeal bitter neurons during feeding events elicits changes in feeding parameters that can suppress intake. Functional dissection experiments indicate that multiple classes of pharyngeal neurons are involved in achieving behavioral avoidance, by virtue of being inhibited or activated by aversive tastants. Tracing second-order pharyngeal circuits reveals two main relay centers for processing pharyngeal taste inputs. Together, our results suggest that the pharynx can control the ingestion of harmful compounds by integrating taste input from different classes of pharyngeal neurons.
Sigma-Aldrich
Strychnine hydrochloride
Sigma-Aldrich
all trans-Retinal, powder, ≥98%
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Goat serum
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
(−)-Lobeline hydrochloride, 98%
Sigma-Aldrich
L-(+)-Tartaric acid, ACS reagent, ≥99.5%
Lobeline hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
DEET, PESTANAL®, analytical standard
Sigma-Aldrich
Theophylline, anhydrous, ≥99%, powder
Sigma-Aldrich
Denatonium benzoate, ≥98%
Sigma-Aldrich
Papaverine hydrochloride, powder
Sigma-Aldrich
Sulforhodamine B, Dye content 75 %
Sigma-Aldrich
Quinine hydrochloride dihydrate, BioReagent, suitable for fluorescence, ≥98.0% (NT)