Merck
  • Metabolism of some "second"- and "fourth"-generation antidepressants: iprindole, viloxazine, bupropion, mianserin, maprotiline, trazodone, nefazodone, and venlafaxine.
Metabolism of some "second"- and "fourth"-generation antidepressants: iprindole, viloxazine, bupropion, mianserin, maprotiline, trazodone, nefazodone, and venlafaxine.
Cellular and molecular neurobiology (1999-06-24)
S Rotzinger, M Bourin, Y Akimoto, R T Coutts, G B Baker
ABSTRACT
1. This review summarizes the major known aspects of the metabolism of second-generation (iprindole, viloxazine, bupropion, mianserin, maprotiline, and trazodone) and fourth-generation (nefazodone and venlafaxine) antidepressants. 2. Discussions about specific enzymes involved and about possible pharmacokinetic drug-drug interactions, particularly as they relate to cytochrome P450 enzymes, are provided.
Sigma-Aldrich
Trazodone hydrochloride, ≥99% (HPLC), powder
Sigma-Aldrich
Maprotiline hydrochloride, >99% (HPLC), powder