Merck
  • Calpain 9 as a therapeutic target in TGFβ-induced mesenchymal transition and fibrosis.
Calpain 9 as a therapeutic target in TGFβ-induced mesenchymal transition and fibrosis.
Science translational medicine (2019-07-19)
David H Kim, James D Beckett, Varun Nagpal, Manuel A Seman-Senderos, Russell A Gould, Tyler J Creamer, Elena Gallo MacFarlane, Yichun Chen, Djahida Bedja, Jonathan T Butcher, Wayne Mitzner, Rosanne Rouf, Shoji Hata, Daniel S Warren, Harry C Dietz
ABSTRACT
Fibrosis is a common pathologic outcome of chronic disease resulting in the replacement of normal tissue parenchyma with a collagen-rich extracellular matrix produced by myofibroblasts. Although the progenitor cell types and cellular programs giving rise to myofibroblasts through mesenchymal transition can vary between tissues and diseases, their contribution to fibrosis initiation, maintenance, and progression is thought to be pervasive. Here, we showed that the ability of transforming growth factor-β (TGFβ) to efficiently induce myofibroblast differentiation of cultured epithelial cells, endothelial cells, or quiescent fibroblasts is dependent on the induced expression and activity of dimeric calpains, a family of non-lysosomal cysteine proteases that regulate a variety of cellular events through posttranslational modification of diverse substrates. siRNA-based gene silencing demonstrated that TGFβ-induced mesenchymal transition of a murine breast epithelial cell line was dependent on induction of expression of calpain 9 (CAPN9), an isoform previously thought to be restricted to the gastrointestinal tract. Mice lacking functional CAPN9 owing to biallelic targeting of Capn9 were viable and fertile but showed overt protection from bleomycin-induced lung fibrosis, carbon tetrachloride-induced liver fibrosis, and angiotensin II-induced cardiac fibrosis and dysfunction. A predicted loss-of-function allele of CAPN9 is common in Southeast Asia, with the frequency of homozygosity matching the prediction of Hardy-Weinberg equilibrium. Together with the highly spatially restricted pattern of CAPN9 expression under physiologic circumstances and the heartiness of the murine knockout, these data provide a strong signature for tolerance of therapeutic strategies for fibrosis aimed at CAPN9 antagonism.
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle - Cy3 antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
SB 431542 hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
CA-074 methyl ester, ≥99% (TLC)
Sigma-Aldrich
Anti-phospho-Smad2 (Ser465/467) Antibody, clone A5S, rabbit monoclonal, culture supernatant, clone A5S, Upstate®
Sigma-Aldrich
Anti-CAPN9 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
2-Aminoethyl diphenylborinate, 97%