Merck
  • Fully rubbery integrated electronics from high effective mobility intrinsically stretchable semiconductors.
Fully rubbery integrated electronics from high effective mobility intrinsically stretchable semiconductors.
Science advances (2019-02-13)
Kyoseung Sim, Zhoulyu Rao, Hae-Jin Kim, Anish Thukral, Hyunseok Shim, Cunjiang Yu
ABSTRACT
An intrinsically stretchable rubbery semiconductor with high mobility is critical to the realization of high-performance stretchable electronics and integrated devices for many applications where large mechanical deformation or stretching is involved. Here, we report fully rubbery integrated electronics from a rubbery semiconductor with a high effective mobility, obtained by introducing metallic carbon nanotubes into a rubbery semiconductor composite. This enhancement in effective carrier mobility is enabled by providing fast paths and, therefore, a shortened carrier transport distance. Transistors and their arrays fully based on intrinsically stretchable electronic materials were developed, and they retained electrical performances without substantial loss when subjected to 50% stretching. Fully rubbery integrated electronics and logic gates were developed, and they also functioned reliably upon mechanical stretching. A rubbery active matrix based elastic tactile sensing skin to map physical touch was demonstrated to illustrate one of the applications.