Merck
  • Epigenetic Alterations Induced by Photothrombotic Stroke in the Rat Cerebral Cortex: Deacetylation of Histone h3, Upregulation of Histone Deacetylases and Histone Acetyltransferases.
Epigenetic Alterations Induced by Photothrombotic Stroke in the Rat Cerebral Cortex: Deacetylation of Histone h3, Upregulation of Histone Deacetylases and Histone Acetyltransferases.
International journal of molecular sciences (2019-06-16)
Svetlana Demyanenko, Anatoly Uzdensky
ABSTRACT
Ischemic penumbra that surrounds a stroke-induced infarction core is potentially salvageable; however, mechanisms of its formation are not well known. Covalent modifications of histones control chromatin conformation, gene expression and protein synthesis. To study epigenetic processes in ischemic penumbra, we used photothrombotic stroke (PTS), a stroke model in which laser irradiation of the rat brain cortex photosensitized by Rose Bengal induces local vessel occlusion. Immunoblotting and immunofluorescence microscopy showed decrease in acetylation of lysine 9 in histone H3 in penumbra at 1, 4 or 24 h after PTS. This was associated with upregulation of histone deacetylases HDAC1 and HDAC2, but not HDAC4, which did not localize in the nuclei. HDAC2 was found in cell nuclei, HDAC4 in the cytoplasm and HDAC1 both in nuclei and cytoplasm. Histone acetyltransferases HAT1 and PCAF (p300/CBP associated factor) that acetylated histone H3 synthesis were also upregulated, but lesser and later. PTS increased localization of HDAC2 and HAT1 in astroglia. Thus, the cell fate in PTS-induced penumbra is determined by the balance between opposite tendencies leading either to histone acetylation and stimulation of gene expression, or to deacetylation and suppression of transcriptional processes and protein biosynthesis. These epigenetic proteins may be the potential targets for anti-stroke therapy.
Sigma-Aldrich
Anti-Rabbit IgG (H+L), highly cross-adsorbed, CF 488A antibody produced in goat, ~2 mg/mL, affinity isolated antibody
Sigma-Aldrich
Anti-HAT antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Histone H3 (Acetyl-Lys9) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-HDAC4 (Ab-632) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Histone Deacetylase 2 (HDAC2) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Histone Deacetylase 1 (HDAC1) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Neuron-Specific Enolase (NSE), Mouse monoclonal, clone NSE-P1, purified from hybridoma cell culture