Merck
  • Identification and action of phenolic compounds of Jatobá-do-cerrado (Hymenaea stignocarpa Mart.) on α-amylase and α-glucosidase activities and flour effect on glycemic response and nutritional quality of breads.
Identification and action of phenolic compounds of Jatobá-do-cerrado (Hymenaea stignocarpa Mart.) on α-amylase and α-glucosidase activities and flour effect on glycemic response and nutritional quality of breads.
Food research international (Ottawa, Ont.) (2019-02-06)
Cintia Pereira da Silva, Rosana Aparecida Manólio Soares-Freitas, Geni Rodrigues Sampaio, Millena Cristina Barros Santos, Talita Pimenta do Nascimento, Luiz Claudio Cameron, Mariana Simões Larraz Ferreira, José Alfredo Gomes Arêas
ABSTRACT
This study investigated the phenolic profile of jatobá-do-cerrado (Hymenaea stignocarpa Mart.) extracts submitted to in vitro digestion, the inhibition of α-amylase and α-glucosidase activities; and the effect of jatobá-do-cerrado flour addition on the nutritional quality, glycemic index (GI) and acceptability of breads. The phenolic composition was determined by UPLC-MS. Bread formulations were developed adding the jatobá-do-cerrado flour at 10, 20 and 30% (w/w) to replace wheat flour. Phenolic compounds of biological relevance such as caffeic acid, kaempferol, quercetin-3-rutinoside and quercetin-3-rhamnoside were present in jatobá-do-cerrado. The phenolic extracts after in vitro digestion significantly inhibited the α-amylase and α-glucosidase activities. Breads with 20% jatobá-do-cerrado flour addition promoted significant reduction (22%) in GI from 70 (control) to 54.3, while 30% addition presented GI of 57.4 and 10% addition a GI of 62.5. The replacement of refined wheat flour by jatobá-do-cerrado flour reduced glycemic response of breads in a non-dose dependent manner. The dietary fibers and the phenolic compounds of jatobá-do-cerrado exerted a synergetic modulation of glucose metabolism by inhibiting sugar metabolic enzymes and glucose absorption. Thus, jatobá-do-cerrado can be included in the diet of healthy individuals and chronic diseases patients, such as diabetics, as an alternative to improve glycemic control.