Merck
  • Epigenetic Modification Is Regulated by the Interaction of Influenza A Virus Nonstructural Protein 1 with the De Novo DNA Methyltransferase DNMT3B and Subsequent Transport to the Cytoplasm for K48-Linked Polyubiquitination.
Epigenetic Modification Is Regulated by the Interaction of Influenza A Virus Nonstructural Protein 1 with the De Novo DNA Methyltransferase DNMT3B and Subsequent Transport to the Cytoplasm for K48-Linked Polyubiquitination.
Journal of virology (2019-01-18)
Shi Liu, Li Liu, Gang Xu, Zhongying Cao, Qing Wang, Shun Li, Nanfang Peng, Jingchuan Yin, Haisheng Yu, Mengqi Li, Zhangchuan Xia, Li Zhou, Yong Lin, Xueyu Wang, Qian Li, Chengliang Zhu, Xuecheng Yang, Jun Wang, Yinglong She, Mengji Lu, Ying Zhu
ABSTRACT
The influenza virus nonstructural protein 1 (NS1) is a nonstructural protein that plays a major role in antagonizing host interferon responses during infection. However, a clear role for the NS1 protein in epigenetic modification has not been established. In this study, NS1 was found to regulate the expression of some key regulators of JAK-STAT signaling by inhibiting the DNA methylation of their promoters. Furthermore, DNA methyltransferase 3B (DNMT3B) is responsible for this process. Upon investigating the mechanisms underlying this event, NS1 was found to interact with DNMT3B but not DNMT3A, leading to the dissociation of DNMT3B from the promoters of the corresponding genes. In addition, the interaction between NS1 and DNMT3B changed the localization of DNMT3B from the nucleus to the cytosol, resulting in K48-linked ubiquitination and degradation of DNMT3B in the cytosol. We conclude that NS1 interacts with DNMT3B and changes its localization to mediate K48-linked polyubiquitination, subsequently contributing to the modulation of the expression of JAK-STAT signaling suppressors.IMPORTANCE The nonstructural protein 1 (NS1) of the influenza A virus (IAV) is a multifunctional protein that counters cellular antiviral activities and is a virulence factor. However, the involvement of NS1 in DNA methylation during IAV infection has not been established. Here, we reveal that the NS1 protein binds the cellular DNMT3B DNA methyltransferase, thereby inhibiting the methylation of the promoters of genes encoding suppressors of JAK-STAT signaling. As a result, these suppressor genes are induced, and JAK-STAT signaling is inhibited. Furthermore, we demonstrate that the NS1 protein transports DNMT3B to the cytoplasm for ubiquitination and degradation. Thus, we identify the NS1 protein as a potential trigger of the epigenetic deregulation of JAK-STAT signaling suppressors and illustrate a novel mechanism underlying the regulation of host immunity during IAV infection.