Merck
  • Stem cell proliferation is induced by apoptotic bodies from dying cells during epithelial tissue maintenance.
Stem cell proliferation is induced by apoptotic bodies from dying cells during epithelial tissue maintenance.
Nature communications (2019-03-07)
Courtney K Brock, Stephen T Wallin, Oscar E Ruiz, Krystin M Samms, Amrita Mandal, Elizabeth A Sumner, George T Eisenhoffer
ABSTRACT
Epithelial tissues require the removal and replacement of damaged cells to sustain a functional barrier. Dying cells provide instructive cues that can influence surrounding cells to proliferate, but how these signals are transmitted to their healthy neighbors to control cellular behaviors during tissue homeostasis remains poorly understood. Here we show that dying stem cells facilitate communication with adjacent stem cells by caspase-dependent production of Wnt8a-containing apoptotic bodies to drive cellular turnover in living epithelia. Basal stem cells engulf apoptotic bodies, activate Wnt signaling, and are stimulated to divide to maintain tissue-wide cell numbers. Inhibition of either cell death or Wnt signaling eliminated the apoptosis-induced cell division, while overexpression of Wnt8a signaling combined with induced cell death led to an expansion of the stem cell population. We conclude that ingestion of apoptotic bodies represents a regulatory mechanism linking death and division to maintain overall stem cell numbers and epithelial tissue homeostasis.
Sigma-Aldrich
IWR-1, ≥98% (HPLC)
Sigma-Aldrich
Caspase-3 Inhibitor II, The Caspase-3 Inhibitor II, also referenced under CAS 210344-95-9, controls the biological activity of Caspase-3. This small molecule/inhibitor is primarily used for Cancer applications.
Sigma-Aldrich
Apoptosis Inhibitor II, NS3694, The Apoptosis Inhibitor II, NS3694, also referenced under CAS 426834-38-0, controls the biological activity of Apoptosis. This small molecule/inhibitor is primarily used for Cancer applications.
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse